Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014390.2 | NP_055205.2 | 4, 5 | 646 | ||
XM_017011987.1 | XP_016867476.1 | 4, 5 | 675 | |||
GenBank mRNA | AB209510.1 | - | 4, 5 | 584 | ||
AK096583.1 | - | 5, 6 | 571 | |||
AK304181.1 | - | 4, 5 | 538 | |||
AK312313.1 | - | 4, 5 | 600 | |||
BC017180.2 | - | 4, 5 | 602 | 3345037 | ||
BT009785.1 | - | 4, 5 | 420 | |||
U22055.1 | - | 4, 5 | 612 |