Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001527.3 | NP_001518.3 | 2 | 411 | ||
NR_033441.1 | 3 | 439 | ||||
NR_073443.1 | 2 | 317 | ||||
XM_011535788.1 | XP_011534090.1 | 3 | 163 | |||
XM_017010799.1 | XP_016866288.1 | 3 | 385 | |||
GenBank mRNA | AK092156.1 | - | 2 | 317 | ||
AK097376.1 | - | 3 | 373 | |||
AK296856.1 | - | 2 | 411 | |||
BC031055.1 | - | 2 | 375 | |||
BP201509.1 | - | 2 | 406 | |||
U31814.1 | - | 2 | 328 |