Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKCI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002740.5 | NP_002731.4 | 13 | 1478 | ||
XM_017006860.1 | XP_016862349.1 | 12 | 1385 | |||
GenBank mRNA | AB451443.1 | - | 13 | 1213 | ||
AK315342.1 | - | 13 | 1406 | |||
BC022016.2 | - | 13 | 1473 | 4823853 | ||
BC042405.1 | - | 1 | 244 | |||
L18964.1 | - | 13 | 1477 | |||
L33881.1 | - | 13 | 1417 |