Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003339.2 | NP_003330.1 | 7 | 1730 | ||
NM_181838.1 | NP_862821.1 | 8 | 1894 | |||
XM_017009820.1 | XP_016865309.1 | 10 | 2101 | |||
GenBank mRNA | AF317220.1 | - | 5 | 1038 | ||
AK001311.1 | - | 7 | 1503 | |||
AK001428.1 | - | 7 | 1557 | |||
AY651263.1 | - | 7 | 1486 | |||
BC019052.2 | - | 7 | 1514 | |||
BC033349.1 | - | 7 | 1730 | 4827210 |