Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003345.4 | NP_003336.1 | 6 | 560 | ||
NM_194259.2 | NP_919235.1 | 7 | 817 | |||
NM_194260.2 | NP_919236.1 | 6 | 680 | |||
NM_194261.2 | NP_919237.1 | 6 | 532 | |||
XM_005255540.2 | XP_005255597.1 | 6 | 551 | |||
XM_005255541.4 | XP_005255598.1 | 6 | 957 | |||
XM_011522644.2 | XP_011520946.1 | 7 | 797 | |||
XM_011522645.1 | XP_011520947.1 | 6 | 662 | |||
XM_017023640.1 | XP_016879129.1 | 6 | 957 | |||
GenBank mRNA | AB208988.1 | - | 6 | 641 | ||
AJ002385.1 | - | 6 | 480 | |||
AK024172.1 | - | 1 | 1252 | |||
AK291118.1 | - | 6 | 551 | |||
BC000427.2 | - | 6 | 464 | 2819764 | ||
BC004429.1 | - | 6 | 464 | 2819764 | ||
BC051289.1 | - | 6 | 494 | |||
BQ230918.1 | - | 1 | 50 | |||
BT006932.1 | - | 5 | 402 | |||
BT007010.1 | - | 5 | 402 | |||
BU146357.1 | - | 2 | 133 | |||
BU543455.1 | - | 6 | 552 | |||
CD249028.1 | - | 6 | 501 | |||
CX788607.1 | - | 6 | 636 | |||
D45050.1 | - | 6 | 519 | |||
U29092.1 | - | 5 | 426 | |||
U31882.1 | - | 6 | 495 | |||
U31933.1 | - | 6 | 520 | |||
U38785.1 | - | 6 | 494 | |||
U45328.1 | - | 6 | 525 | |||
U66818.1 | - | 6 | 489 | |||
U66867.1 | - | 6 | 1208 | |||
X96427.1 | - | 6 | 525 |