Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320595.1 | NP_001307524.1 | 8 | 1223 | ||
NM_001320596.1 | NP_001307525.1 | 8 | 1112 | |||
NM_001320597.1 | NP_001307526.1 | 7 | 963 | |||
NM_004396.4 | NP_004387.1 | 7 | 1198 | |||
GenBank mRNA | AB209919.1 | - | 7 | 950 | ||
AB451257.1 | - | 7 | 796 | |||
AB451382.1 | - | 7 | 796 | |||
AK297192.1 | - | 6 | 728 | |||
AK297753.1 | - | 8 | 933 | |||
AK297905.1 | - | 6 | 756 | |||
AK310318.1 | - | 7 | 842 | |||
BC016027.1 | - | 7 | 937 | 3528578 | ||
BT006943.1 | - | 7 | 796 | |||
BX571764.1 | - | 7 | 967 | |||
X15729.1 | - | 7 | 963 | |||
X52104.1 | - | 7 | 971 |