Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257191.2 | NP_001244120.1 | 13, 14 | 1825 | ||
NM_004397.5 | NP_004388.2 | 13, 14 | 1897 | |||
XM_005271417.3 | XP_005271474.1 | 13, 14 | 1842 | |||
XM_011542644.1 | XP_011540946.1 | 12, 13 | 1768 | |||
XM_011542645.1 | XP_011540947.1 | 12, 13 | 1750 | |||
XM_017017251.1 | XP_016872740.1 | 12, 13 | 1713 | |||
GenBank mRNA | AK093657.1 | - | 5, 6 | 1067 | ||
BC065007.1 | - | 12, 13 | 1756 | 6163439 | ||
D17532.1 | - | 13, 14 | 1770 | |||
Z11685.1 | - | 13, 14 | 1797 |