Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004667.5 | NP_004658.3 | 22 | 3410 | ||
XM_005268276.4 | XP_005268333.1 | 21 | 3540 | |||
XM_006720726.3 | XP_006720789.1 | 22 | 3431 | |||
XM_006720727.3 | XP_006720790.1 | 20 | 3173 | |||
XM_011522131.2 | XP_011520433.1 | 20 | 3197 | |||
XM_017022695.1 | XP_016878184.1 | 21 | 3357 | |||
XM_017022696.1 | XP_016878185.1 | 21 | 3300 | |||
GenBank mRNA | AF071172.3 | - | 22 | 3362 |