Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015053.1 | NP_001015053.1 | 7, 8 | 1115 | ||
NM_005474.4 | NP_005465.2 | 7, 8 | 1112 | |||
XM_005256905.1 | XP_005256962.1 | 7, 8 | 1110 | |||
XM_005256906.4 | XP_005256963.1 | 7, 8 | 1071 | |||
XM_011524149.1 | XP_011522451.1 | 7, 8 | 1086 | |||
XM_011524150.1 | XP_011522452.1 | 6, 7 | 862 | |||
XM_017023988.1 | XP_016879477.1 | 9, 10 | 1224 | |||
XM_017023989.1 | XP_016879478.1 | 9, 10 | 1190 | |||
XM_017023990.1 | XP_016879479.1 | 9, 10 | 1215 | |||
XM_017023991.1 | XP_016879480.1 | 9, 10 | 1221 | |||
XM_017023992.1 | XP_016879481.1 | 9, 10 | 1225 | |||
XM_017023993.1 | XP_016879482.1 | 9, 10 | 1197 | |||
XM_017023994.1 | XP_016879483.1 | 5, 6 | 1178 | |||
XM_017023995.1 | XP_016879484.1 | 7, 8 | 1080 | |||
XM_017023996.1 | XP_016879485.1 | 8, 9 | 1287 | |||
GenBank mRNA | AB011172.2 | - | 7, 8 | 1084 | ||
AF039691.1 | - | 2, 3 | 217 | |||
AF132608.1 | - | 6, 7 | 780 | |||
AK294760.1 | - | 7, 8 | 1080 | |||
BC013140.1 | - | 4, 5 | 562 | |||
BC051824.1 | - | 7, 8 | 991 | 6043491 |