Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETMAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243723.1 | NP_001230652.1 | 2 | 256 | ||
NM_001276325.1 | NP_001263254.1 | 2 | 256 | |||
NM_001320677.1 | NP_001307606.1 | 2 | 256 | |||
NM_006515.3 | NP_006506.3 | 2 | 256 | |||
XM_006713294.3 | XP_006713357.1 | 2 | 318 | |||
XM_017007032.1 | XP_016862521.1 | 2 | 319 | |||
GenBank mRNA | AF054989.1 | - | 2 | 182 | ||
AK222734.1 | - | 2 | 192 | |||
AK297865.1 | - | 2 | 191 | |||
AK302296.1 | - | 2 | 190 | |||
AK307537.1 | - | 2 | 256 | |||
AK311418.1 | - | 2 | 289 | |||
AY952295.1 | - | 2 | 161 | |||
BC011635.2 | - | 2 | 182 | 4080032 | ||
BG479260.1 | - | 2 | 271 | |||
DC357038.1 | - | 2 | 256 | |||
U80776.1 | - | 2 | 173 |