Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289413.1 | NP_001276342.1 | 8 | 1212 | ||
NM_001318833.1 | NP_001305762.1 | 9 | 1296 | |||
NM_006709.4 | NP_006700.3 | 9 | 1067 | |||
NM_025256.6 | NP_079532.5 | 9 | 1067 | |||
XM_005248824.3 | XP_005248881.1 | 8 | 1208 | |||
XM_006714974.1 | XP_006715037.1 | 8 | 1212 | |||
XM_006714975.1 | XP_006715038.1 | 8 | 1212 | |||
XM_006714976.1 | XP_006715039.1 | 9 | 1050 | |||
XM_017010202.1 | XP_016865691.1 | 9 | 1050 | |||
GenBank mRNA | AB209433.2 | - | 8 | 1129 | ||
AJ315532.1 | - | 9 | 1037 | |||
AK302904.1 | - | 8 | 1212 | |||
BC002686.1 | - | 6 | 537 | |||
BC009351.1 | - | 9 | 1034 | |||
BC018718.1 | - | 9 | 1025 | 4123580 | ||
BC020970.1 | - | 8 | 1002 | |||
X69838.1 | - | 5 | 457 |