Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL20RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278722.1 | NP_001265651.1 | 4 | 564 | ||
NM_001278723.1 | NP_001265652.1 | 3 | 704 | |||
NM_014432.3 | NP_055247.3 | 4 | 883 | |||
XM_011535904.2 | XP_011534206.1 | 3 | 391 | |||
XM_017010954.1 | XP_016866443.1 | 4 | 1072 | |||
XM_017010955.1 | XP_016866444.1 | 4 | 532 | |||
GenBank mRNA | AF184971.1 | - | 4 | 667 | ||
AK297121.1 | - | 4 | 526 | |||
AK315293.1 | - | 4 | 671 | |||
AY358883.1 | - | 3 | 218 | |||
BC113574.1 | - | 4 | 761 | 8322626 | ||
BC113602.1 | - | 4 | 761 | 8322654 |