Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014433.2 | NP_055248.1 | 3 | 397 | ||
XM_011530149.2 | XP_011528451.1 | 3 | 436 | |||
XM_011530150.1 | XP_011528452.1 | 3 | 436 | |||
XM_011530151.2 | XP_011528453.1 | 3 | 599 | |||
XM_011530152.2 | XP_011528454.1 | 3 | 527 | |||
XM_011530154.2 | XP_011528456.1 | 2 | 142 | |||
XM_017028774.1 | XP_016884263.1 | 3 | 422 | |||
GenBank mRNA | AF133587.1 | - | 3 | 397 | ||
AK299530.1 | - | 3 | 355 | |||
AK310327.1 | - | 3 | 436 | |||
BC008986.1 | - | 3 | 365 | 4339021 | ||
CR456567.1 | - | 2 | 297 |