Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282659.1 | NP_001269588.1 | 10 | 1878 | ||
NM_017944.3 | NP_060414.3 | 9 | 1674 | |||
XM_005252997.1 | XP_005253054.1 | 11 | 1938 | |||
XM_011520196.1 | XP_011518498.1 | 3 | 1053 | |||
XM_017017953.1 | XP_016873442.1 | 11 | 1994 | |||
XM_017017954.1 | XP_016873443.1 | 3 | 2201 | |||
GenBank mRNA | AK027283.1 | - | 786 | |||
AK027362.1 | - | 9 | 1079 | |||
AK092290.1 | - | 11 | 1295 | |||
AK127264.1 | - | 10 | 1674 | |||
AK309240.1 | - | 10 | 1342 | |||
DA490237.1 | - | 2 | 95 |