Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166418.1 | NP_001159890.1 | 6 | 837 | ||
NM_018486.2 | NP_060956.1 | 8 | 1110 | |||
NR_051952.1 | 7 | 968 | ||||
XM_011530986.2 | XP_011529288.2 | 8 | 1105 | |||
XM_011530987.2 | XP_011529289.2 | 8 | 1105 | |||
XM_017029640.1 | XP_016885129.1 | 7 | 1027 | |||
XM_017029641.1 | XP_016885130.1 | 7 | 1027 | |||
XM_017029642.1 | XP_016885131.1 | 6 | 830 | |||
XM_017029643.1 | XP_016885132.1 | 5 | 805 | |||
XM_017029644.1 | XP_016885133.1 | 5 | 754 | |||
XM_017029645.1 | XP_016885134.1 | 5 | 805 | |||
XM_017029646.1 | XP_016885135.1 | 7 | 654 | |||
GenBank mRNA | AF230097.1 | - | 8 | 823 | ||
AF245664.1 | - | 8 | 777 | |||
AJ277724.1 | - | 8 | 810 | |||
AK294903.1 | - | 6 | 551 | |||
AK296641.1 | - | 6 | 787 | |||
AK298767.1 | - | 7 | 708 | |||
AY429539.1 | - | 7 | 654 | |||
BC050433.1 | - | 8 | 801 | 5761745 |