Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASH1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018489.2 | NP_060959.2 | 3 | 5284 | ||
XM_005245337.4 | XP_005245394.1 | 3 | 5284 | |||
XM_006711450.3 | XP_006711513.1 | 3 | 4975 | |||
XM_006711451.3 | XP_006711514.1 | 4 | 5062 | |||
XM_006711452.3 | XP_006711515.1 | 3 | 5284 | |||
XM_011509769.2 | XP_011508071.1 | 3 | 5284 | |||
XM_011509770.2 | XP_011508072.1 | 1 | 564 | |||
XM_017001784.1 | XP_016857273.1 | 3 | 5061 | |||
XM_017001785.1 | XP_016857274.1 | 3 | 4991 | |||
XM_017001786.1 | XP_016857275.1 | 3 | 5284 | |||
XM_017001787.1 | XP_016857276.1 | 3 | 5284 | |||
GenBank mRNA | AF257305.1 | - | 3 | 5113 |