Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033604.1 | NP_001028776.1 | 4, 5 | 849 | ||
NM_001033605.1 | NP_001028777.1 | 4, 5 | 849 | |||
NM_014451.3 | NP_055266.2 | 4, 5 | 849 | |||
NM_198428.2 | NP_940820.1 | 4, 5 | 849 | |||
XM_005249700.3 | XP_005249757.1 | 4, 5 | 857 | |||
XM_005249701.2 | XP_005249758.1 | 4, 5 | 857 | |||
XM_011515264.2 | XP_011513566.1 | 4, 5 | 857 | |||
XM_011515265.2 | XP_011513567.1 | 4, 5 | 656 | |||
XM_011515266.2 | XP_011513568.1 | 4, 5 | 857 | |||
XM_011515267.2 | XP_011513569.1 | 4, 5 | 857 | |||
XM_011515268.1 | XP_011513570.1 | 4, 5 | 857 | |||
XM_011515269.2 | XP_011513571.1 | 4, 5 | 840 | |||
XM_011515270.2 | XP_011513572.1 | 4, 5 | 857 | |||
XM_017011990.1 | XP_016867479.1 | 4, 5 | 563 | |||
XM_017011991.1 | XP_016867480.1 | 4, 5 | 565 | |||
XM_017011992.1 | XP_016867481.1 | 4, 5 | 840 | |||
XM_017011993.1 | XP_016867482.1 | 4, 5 | 840 | |||
XM_017011994.1 | XP_016867483.1 | 4, 5 | 857 | |||
GenBank mRNA | AF095771.1 | - | 3, 4 | 330 | ||
AK057660.1 | - | 4, 5 | 827 | |||
AK314627.1 | - | 4, 5 | 690 | |||
BC032715.1 | - | 3, 4 | 271 | |||
BC103831.2 | - | 4, 5 | 469 | 40001794 | ||
BX647385.1 | - | 4, 5 | 827 |