Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETDB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320699.1 | NP_001307628.1 | 3, 4 | 768 | ||
NM_031915.2 | NP_114121.2 | 4, 5 | 1125 | |||
GenBank mRNA | AF334407.1 | - | 4, 5 | 1044 |