Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETDB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160308.2 | NP_001153780.1 | 12 | 2639 | ||
NM_001320699.1 | NP_001307628.1 | 12 | 2318 | |||
NM_031915.2 | NP_114121.2 | 13 | 2675 | |||
GenBank mRNA | AB209349.1 | - | 5 | 4879 | ||
AF086317.1 | - | 1 | 23 | |||
AF334407.1 | - | 13 | 2594 | |||
AK307593.1 | - | 11 | 1988 | |||
AK308220.1 | - | 12 | 2556 | |||
AL831937.1 | - | 6 | 775 | |||
BC017078.1 | - | 4 | 477 | |||
BC047434.2 | - | 11 | 2075 | 5266911 |