Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032233.2 | NP_115609.2 | 8 | 936 | ||
NM_199123.1 | NP_954574.1 | 8 | 936 | |||
XM_005268127.3 | XP_005268184.1 | 7 | 789 | |||
XM_011537231.2 | XP_011535533.2 | 8 | 888 | |||
XM_011537232.2 | XP_011535534.1 | 8 | 2066 | |||
XM_011537234.1 | XP_011535536.1 | 7 | 656 | |||
XM_011537235.1 | XP_011535537.1 | 8 | 888 | |||
XM_017021699.1 | XP_016877188.1 | 7 | 784 | |||
XM_017021700.1 | XP_016877189.1 | 8 | 972 | |||
GenBank mRNA | AK026680.1 | - | 7 | 690 | ||
AK302882.1 | - | 8 | 2279 | |||
AL359581.2 | - | 3 | 126 | |||
BC009054.1 | - | 5 | 562 | |||
BC066931.1 | - | 8 | 837 | |||
BC120967.1 | - | 8 | 791 | |||
BC120968.2 | - | 8 | 791 | |||
BC127624.1 | - | 8 | 791 | |||
BC127625.1 | - | 8 | 791 | |||
BC142995.1 | - | 8 | 948 | 8860450 | ||
BC148251.1 | - | 7 | 787 | |||
BG194029.1 | - | 96 | ||||
BX161441.1 | - | 8 | 835 | |||
BX161471.1 | - | 8 | 926 |