Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290325.1 | NP_001277254.1 | 9 | 3449 | ||
NM_001290326.1 | NP_001277255.1 | 10 | 3565 | |||
NM_032557.6 | NP_115946.2 | 9 | 3449 | |||
XM_011532360.2 | XP_011530662.1 | 8 | 2835 | |||
GenBank mRNA | AB067478.1 | - | 8 | 2130 | ||
AF211481.1 | - | 1 | 1022 | |||
AK057992.1 | - | 6 | 1973 | |||
AK091712.1 | - | 2 | 1223 | |||
AK093289.1 | - | 1 | 499 | |||
AK093754.1 | - | 10 | 2788 | |||
AK126943.1 | - | 10 | 3277 | |||
AL833976.1 | - | 9 | 3421 | |||
BC039115.1 | - | 9 | 3449 | 5258804 | ||
BC068975.1 | - | 9 | 3396 | 6067501 | ||
BG575871.1 | - | 1 | 558 |