Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301175.1 | NP_001288104.1 | 16 | 2151 | ||
NM_032663.4 | NP_116052.2 | 13 | 1777 | |||
XM_005253962.3 | XP_005254019.1 | 13 | 1760 | |||
XM_005253965.4 | XP_005254022.1 | 12 | 1588 | |||
XM_006719653.3 | XP_006719716.1 | 14 | 1850 | |||
XM_011538894.2 | XP_011537196.1 | 11 | 1481 | |||
XM_017020048.1 | XP_016875537.1 | 13 | 1883 | |||
XM_017020049.1 | XP_016875538.1 | 13 | 1871 | |||
XM_017020050.1 | XP_016875539.1 | 13 | 1983 | |||
XM_017020051.1 | XP_016875540.1 | 14 | 1839 | |||
XM_017020052.1 | XP_016875541.1 | 13 | 1885 | |||
XM_017020053.1 | XP_016875542.1 | 12 | 1615 | |||
XM_017020054.1 | XP_016875543.1 | 11 | 1490 | |||
GenBank mRNA | AK027820.1 | - | 13 | 1742 | ||
AK097830.1 | - | 16 | 2151 | |||
AL834278.1 | - | 4 | 940 | |||
BC004868.2 | - | 13 | 1767 | 3833242 | ||
BC022094.2 | - | 11 | 1319 |