Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMYD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052928.2 | NP_443160.2 | 11 | 3468 | ||
XM_017024121.1 | XP_016879610.1 | 12 | 3713 | |||
GenBank mRNA | AL137345.1 | - | 1 | 516 | ||
BC035077.2 | - | 11 | 3387 | 5260399 |