Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AURKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323303.1 | NP_001310232.1 | 7 | 1179 | ||
NM_001323304.1 | NP_001310233.1 | 7 | 942 | |||
NM_001323305.1 | NP_001310234.1 | 8 | 1052 | |||
NM_003600.3 | NP_003591.2 | 6 | 1069 | |||
NM_198433.2 | NP_940835.1 | 8 | 1277 | |||
NM_198434.2 | NP_940836.1 | 7 | 968 | |||
NM_198435.2 | NP_940837.1 | 6 | 858 | |||
NM_198436.2 | NP_940838.1 | 7 | 954 | |||
NM_198437.2 | NP_940839.1 | 6 | 844 | |||
XM_017028034.1 | XP_016883523.1 | 8 | 1399 | |||
XM_017028035.1 | XP_016883524.1 | 6 | 759 | |||
GenBank mRNA | AF008551.1 | - | 5 | 669 | ||
AF011468.1 | - | 6 | 925 | |||
AK301769.1 | - | 6 | 531 | |||
AK312776.1 | - | 6 | 736 | |||
BC001280.1 | - | 7 | 801 | 3456017 | ||
BC002499.2 | - | 6 | 797 | 3051177 | ||
BC006423.1 | - | 6 | 694 | 3532438 | ||
BC027464.1 | - | 7 | 836 | 4449474 | ||
BM458030.1 | - | 7 | 869 | |||
BQ435059.1 | - | 6 | 690 | |||
BU171303.1 | - | 6 | 791 | |||
D84212.1 | - | 7 | 835 |