Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AURKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323303.1 | NP_001310232.1 | 10 | 1603 | ||
NM_001323304.1 | NP_001310233.1 | 10 | 1366 | |||
NM_001323305.1 | NP_001310234.1 | 11 | 1476 | |||
NM_003600.3 | NP_003591.2 | 9 | 1493 | |||
NM_198433.2 | NP_940835.1 | 11 | 1701 | |||
NM_198434.2 | NP_940836.1 | 10 | 1392 | |||
NM_198435.2 | NP_940837.1 | 9 | 1282 | |||
NM_198436.2 | NP_940838.1 | 10 | 1378 | |||
NM_198437.2 | NP_940839.1 | 9 | 1268 | |||
XM_017028034.1 | XP_016883523.1 | 11 | 1823 | |||
XM_017028035.1 | XP_016883524.1 | 9 | 1183 | |||
GenBank mRNA | AF008551.1 | - | 8 | 1093 | ||
AF011468.1 | - | 9 | 1349 | |||
AK301769.1 | - | 9 | 955 | |||
AK312776.1 | - | 9 | 1160 | |||
BC001280.1 | - | 10 | 1225 | 3456017 | ||
BC002499.2 | - | 9 | 1221 | 3051177 | ||
BC006423.1 | - | 9 | 1118 | 3532438 | ||
BC027464.1 | - | 10 | 1260 | 4449474 | ||
D84212.1 | - | 10 | 1259 |
Gene Symbol: VWA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011539757.2 | XP_011538059.1 | 1 | 72 |