Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EZH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321079.1 | NP_001308008.1 | 14, 15 | 1758 | ||
NM_001321081.1 | NP_001308010.1 | 15, 16 | 1822 | |||
NM_001321082.1 | NP_001308011.1 | 14, 15 | 1729 | |||
NM_001991.4 | NP_001982.2 | 15, 16 | 1849 | |||
XM_005257145.2 | XP_005257202.1 | 13, 14 | 1604 | |||
XM_011524517.2 | XP_011522819.1 | 14, 15 | 1697 | |||
XM_017024350.1 | XP_016879839.1 | 14, 15 | 1676 | |||
XM_017024351.1 | XP_016879840.1 | 13, 14 | 1583 | |||
XM_017024352.1 | XP_016879841.1 | 12, 13 | 1434 | |||
GenBank mRNA | AB002386.1 | - | 14, 15 | 1761 | ||
AB004818.1 | - | 14, 15 | 1764 | |||
AK295626.1 | - | 14, 15 | 1711 | |||
AK295853.1 | - | 13, 14 | 1549 | |||
AK299887.1 | - | 14, 15 | 1664 | |||
AK304835.1 | - | 14, 15 | 1662 | |||
AK309121.1 | - | 14, 15 | 1669 | |||
AK310399.1 | - | 13, 14 | 1541 | |||
AK310556.1 | - | 7, 8 | 893 | |||
BC015882.1 | - | 15, 16 | 1783 | 3908776 | ||
BT009782.1 | - | 13, 14 | 1661 | |||
U50315.1 | - | 14, 15 | 1767 |