Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202489.1 | NP_001189418.1 | 4 | 410 | ||
NM_003343.5 | NP_003334.2 | 6 | 492 | |||
NM_182688.2 | NP_872630.1 | 7 | 607 | |||
XM_017028469.1 | XP_016883958.1 | 6 | 451 | |||
GenBank mRNA | AF032456.1 | - | 6 | 459 | ||
AK075143.1 | - | 2 | 633 | |||
AK122700.1 | - | 2 | 1909 | |||
AK290629.1 | - | 6 | 492 | |||
AL834129.1 | - | 2 | 1224 | |||
BC001738.1 | - | 6 | 454 | 3536853 | ||
BC008351.1 | - | 6 | 452 | 3536171 | ||
BC011569.1 | - | 6 | 464 | 4548763 | ||
BG702453.1 | - | 6 | 483 | |||
BT006914.1 | - | 6 | 404 | |||
DA710059.1 | - | 4 | 401 |