Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUVBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319084.1 | NP_001306013.1 | 10 | 1426 | ||
NM_003707.2 | NP_003698.1 | 11 | 1459 | |||
XM_005247841.3 | XP_005247898.1 | 11 | 1380 | |||
GenBank mRNA | AB012122.1 | - | 11 | 1408 | ||
AB451224.1 | - | 11 | 1355 | |||
AB451347.1 | - | 11 | 1355 | |||
AF070735.1 | - | 11 | 1431 | |||
AF099084.1 | - | 11 | 1419 | |||
AJ010058.1 | - | 11 | 1431 | |||
AK222563.1 | - | 11 | 1440 | |||
AK222575.1 | - | 11 | 1431 | |||
AK312290.1 | - | 11 | 1455 | |||
BC002993.1 | - | 11 | 1426 | 2823568 | ||
BC012886.1 | - | 11 | 1388 | 3876400 | ||
BT007057.1 | - | 11 | 1355 | |||
DQ469310.1 | - | 10 | 1263 | |||
Y18418.1 | - | 11 | 1451 |