Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014331.3 | NP_055146.1 | 12 | 2498 | ||
GenBank mRNA | AJ277882.1 | - | 12 | 2218 |