Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033054.2 | NP_001028226.1 | 5 | 1614 | ||
NM_001033055.2 | NP_001028227.1 | 5 | 1623 | |||
NM_001285399.2 | NP_001272328.1 | 6 | 1767 | |||
NM_001285400.2 | NP_001272329.1 | 6 | 1737 | |||
NM_001285401.2 | NP_001272330.1 | 6 | 1731 | |||
NM_001285402.1 | NP_001272331.1 | 6 | 1794 | |||
NM_014336.4 | NP_055151.3 | 6 | 1803 | |||
GenBank mRNA | AB593047.1 | - | 6 | 1781 | ||
AB593048.1 | - | 6 | 1792 | |||
AB593049.1 | - | 6 | 1746 | |||
AB593052.1 | - | 6 | 1724 | |||
AB593053.1 | - | 5 | 1580 | |||
AB593055.1 | - | 6 | 1768 | |||
AF148864.1 | - | 6 | 1891 | |||
AK023970.1 | - | 6 | 1732 | |||
BX537907.1 | - | 6 | 1697 | |||
BX647279.1 | - | 6 | 1768 |