Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278945.1 | NP_001265874.1 | 8 | 1396 | ||
NM_018492.3 | NP_060962.2 | 8 | 1421 | |||
XM_006716368.2 | XP_006716431.1 | 8 | 1345 | |||
GenBank mRNA | AB027249.1 | - | 8 | 1147 | ||
AB027250.1 | - | 8 | 1127 | |||
AF189722.1 | - | 8 | 1111 | |||
AF237709.1 | - | 8 | 1135 | |||
AK027291.1 | - | 8 | 1126 | |||
AK301836.1 | - | 8 | 1356 | |||
BC015191.1 | - | 8 | 1135 | 4082846 | ||
EU794685.1 | - | 8 | 1158 |