Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024757.4 | NP_079033.4 | 16 | 2506 | ||
XM_005266105.4 | XP_005266162.1 | 16 | 2549 | |||
XM_005266110.1 | XP_005266167.1 | 15 | 2421 | |||
XM_006717288.2 | XP_006717351.1 | 16 | 2467 | |||
XM_011519021.2 | XP_011517323.1 | 16 | 3504 | |||
XM_011519022.2 | XP_011517324.1 | 16 | 2959 | |||
XM_011519023.2 | XP_011517325.1 | 16 | 3483 | |||
XM_011519024.2 | XP_011517326.1 | 15 | 3427 | |||
XM_011519025.1 | XP_011517327.1 | 16 | 2669 | |||
XM_011519026.2 | XP_011517328.1 | 15 | 3362 | |||
XM_011519027.2 | XP_011517329.1 | 16 | 3505 | |||
XM_011519029.2 | XP_011517331.1 | 8 | 1094 | |||
XM_011519030.2 | XP_011517332.1 | 2 | 789 | |||
XM_011519031.1 | XP_011517333.1 | 2 | 147 | |||
XM_011519033.2 | XP_011517335.1 | 15 | 3341 | |||
XM_017015134.1 | XP_016870623.1 | 16 | 2937 | |||
XM_017015135.1 | XP_016870624.1 | 16 | 2489 | |||
XM_017015136.1 | XP_016870625.1 | 15 | 2411 | |||
XM_017015137.1 | XP_016870626.1 | 16 | 2648 | |||
XM_017015138.1 | XP_016870627.1 | 15 | 2400 | |||
GenBank mRNA | AB028932.1 | - | 11 | 1326 | ||
AB058779.2 | - | 11 | 1326 | |||
AK022941.1 | - | 15 | 2421 | |||
AY083210.1 | - | 14 | 2376 | |||
BC011608.2 | - | 6 | 723 |