Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015053.1 | NP_001015053.1 | 18, 19 | 2795 | ||
NM_005474.4 | NP_005465.2 | 18, 19 | 2792 | |||
XM_005256905.1 | XP_005256962.1 | 18, 19 | 2790 | |||
XM_005256906.4 | XP_005256963.1 | 18, 19 | 2751 | |||
XM_011524149.1 | XP_011522451.1 | 18, 19 | 2766 | |||
XM_011524150.1 | XP_011522452.1 | 17, 18 | 2542 | |||
XM_017023988.1 | XP_016879477.1 | 20, 21 | 2904 | |||
XM_017023989.1 | XP_016879478.1 | 20, 21 | 2870 | |||
XM_017023990.1 | XP_016879479.1 | 20, 21 | 2895 | |||
XM_017023991.1 | XP_016879480.1 | 20, 21 | 2901 | |||
XM_017023992.1 | XP_016879481.1 | 20, 21 | 2890 | |||
XM_017023993.1 | XP_016879482.1 | 20, 21 | 2877 | |||
XM_017023994.1 | XP_016879483.1 | 16, 17 | 2858 | |||
XM_017023995.1 | XP_016879484.1 | 18, 19 | 2760 | |||
XM_017023996.1 | XP_016879485.1 | 19, 20 | 2967 | |||
GenBank mRNA | AB011172.2 | - | 16, 17 | 2509 | ||
AB177853.1 | - | 8, 9 | 1244 | |||
AF039691.1 | - | 13, 14 | 1897 | |||
AF132608.1 | - | 17, 18 | 2460 | |||
AK027734.1 | - | 4, 5 | 610 | |||
AK075095.1 | - | 9, 10 | 1440 | |||
AK294760.1 | - | 18, 19 | 2760 | |||
BC038423.1 | - | 7, 8 | 1074 | |||
BC051824.1 | - | 18, 19 | 2671 | 6043491 |