Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000729.5 | NP_000720.1 | 5 | 1222 | ||
NM_001174138.2 | NP_001167609.1 | 3 | 455 | |||
GenBank mRNA | AK289716.1 | - | 3 | 397 | ||
BC008283.1 | - | 3 | 455 | 3639109 | ||
BC093055.1 | - | 3 | 397 | 30722619 | ||
BC111026.1 | - | 5 | 1197 | 5199880 | ||
BI756101.1 | - | 3 | 383 | |||
BT006991.1 | - | 2 | 328 | |||
CB961869.1 | - | 3 | 424 | |||
CD250282.1 | - | 3 | 557 | |||
CR542226.1 | - | 2 | 328 |