Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308053.1 | NP_001294982.1 | 10 | 1368 | ||
NM_014353.4 | NP_055168.2 | 9 | 1329 | |||
XM_011522448.1 | XP_011520750.1 | 9 | 1387 | |||
XM_011522450.2 | XP_011520752.1 | 8 | 2146 | |||
GenBank mRNA | AB027137.1 | - | 8 | 1000 | ||
AK308560.1 | - | 9 | 1628 | |||
AY646153.1 | - | 9 | 1346 | |||
BC066913.1 | - | 9 | 1335 | 5262795 |