Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003537.3 | NP_003528.1 | 1 | 335 | ||
GenBank mRNA | BC069133.1 | - | 1 | 335 | 7216989 | |
BC096132.1 | - | 1 | 335 | 40002522 | ||
BC096133.3 | - | 1 | 335 | 40002523 | ||
BC096134.3 | - | 1 | 335 | 40002524 | ||
BC099630.3 | - | 1 | 338 | 40002521 |