Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206997.1 | NP_996880.1 | 1 | 1006 | ||
GenBank mRNA | AY569571.1 | - | 1 | 1006 | ||
BC119779.1 | - | 1 | 1006 | 40109407 | ||
BC122869.1 | - | 1 | 1006 | 40109404 |
Set Membership: |
G-protein coupled receptor |