Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024409.3 | NP_077720.1 | 2 | 452 | ||
XM_011511245.2 | XP_011509547.1 | 2 | 436 | |||
GenBank mRNA | BC069120.1 | - | 2 | 298 | 7216974 | |
BC105065.1 | - | 2 | 377 | 8144068 | ||
BC105067.1 | - | 2 | 377 | 8144070 | ||
DR003533.1 | - | 2 | 460 | |||
HQ419060.1 | - | 1 | 105 | |||
JF420840.1 | - | 1 | 103 |