Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GJC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020435.3 | NP_065168.2 | 2 | 570 | ||
GenBank mRNA | AY285161.1 | - | 1 | 248 | ||
BC035840.1 | - | 2 | 324 | |||
BC089439.1 | - | 2 | 570 | 30558819 |