Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMSB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021103.3 | NP_066926.1 | 3 | 239 | ||
GenBank mRNA | AK130949.1 | - | 3 | 227 | ||
AY453400.1 | - | 2 | 172 | |||
BC016025.1 | - | 3 | 204 | 3909431 | ||
BC016731.1 | - | 3 | 226 | 4095378 | ||
BC107889.1 | - | 3 | 226 | |||
M20259.1 | - | 3 | 204 | |||
S54005.1 | - | 3 | 209 |