Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014366.4 | NP_055181.3 | 13 | 1582 | ||
NM_206825.1 | NP_996561.1 | 13 | 1679 | |||
NM_206826.1 | NP_996562.1 | 13 | 1532 | |||
GenBank mRNA | AF191018.1 | - | 13 | 1579 | ||
AK027514.1 | - | 13 | 1583 | |||
AK027516.1 | - | 13 | 1432 | |||
AK315484.1 | - | 13 | 1495 | |||
AY825265.1 | - | 12 | 1409 | |||
BC001024.2 | - | 13 | 1430 | 3342771 |