Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP8L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291478.1 | NP_001278407.1 | 10 | 1265 | ||
NM_014371.3 | NP_055186.3 | 10 | 1448 | |||
NR_111971.1 | 12 | 1483 | ||||
XM_005259854.3 | XP_005259911.2 | 11 | 1253 | |||
XM_011527916.2 | XP_011526218.1 | 10 | 1355 | |||
GenBank mRNA | AB025905.1 | - | 10 | 1295 | ||
AB451326.1 | - | 9 | 1250 | |||
AB451469.1 | - | 9 | 1250 | |||
AF199414.1 | - | 9 | 1250 | |||
AJ243467.1 | - | 10 | 1296 | |||
AK001594.1 | - | 10 | 1987 | |||
AK094736.1 | - | 12 | 1350 | |||
AK225046.1 | - | 10 | 1315 | |||
AK225291.1 | - | 10 | 1315 | |||
AK295956.1 | - | 10 | 1170 | |||
BC000713.2 | - | 10 | 1300 | 3504388 | ||
BF973758.1 | - | 3 | 256 |