Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025105.2 | NP_001020276.1 | 11 | 2234 | ||
NM_001271741.1 | NP_001258670.1 | 10 | 2114 | |||
NM_001271742.1 | NP_001258671.1 | 10 | 1673 | |||
NM_001892.5 | NP_001883.4 | 10 | 2150 | |||
GenBank mRNA | AF119911.2 | - | 1 | 582 | ||
AF447582.1 | - | 2 | 557 | |||
AK294099.1 | - | 8 | 1225 | |||
BC008717.2 | - | 10 | 1693 | 2822250 | ||
BC020446.2 | - | 1 | 178 | |||
BC040473.1 | - | 1 | 151 | |||
BX647478.1 | - | 2 | 6633 |