Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004267.4 | NP_004258.2 | 2 | 2628 | ||
GenBank mRNA | AB014679.2 | - | 2 | 1937 | ||
AB014680.2 | - | 2 | 1937 | |||
AF083066.1 | - | 1 | 1746 | |||
AK023268.1 | - | 2 | 2578 | |||
BC017499.1 | - | 2 | 2309 | |||
BC036930.1 | - | 2 | 2539 | |||
BC068604.1 | - | 2 | 2578 |