Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006843.2 | NP_006834.2 | 6 | 565 | ||
GenBank mRNA | AK292760.1 | - | 6 | 561 | ||
BC020750.1 | - | 6 | 564 | 4662951 | ||
BG534053.1 | - | 6 | 565 | |||
J05037.1 | - | 6 | 532 |