Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014351.3 | NP_055166.1 | 7 | 955 | ||
XM_011530121.1 | XP_011528423.1 | 4 | 560 | |||
GenBank mRNA | AF115311.1 | - | 7 | 839 | ||
AF176342.1 | - | 7 | 859 | |||
AF188698.1 | - | 7 | 922 | |||
AF251263.1 | - | 7 | 868 | |||
AK091700.1 | - | 1 | 1017 | |||
AK294622.1 | - | 4 | 552 | |||
AK295399.1 | - | 7 | 788 | |||
AK313048.1 | - | 7 | 880 | |||
BC022459.1 | - | 7 | 848 | 4795827 | ||
BC028171.1 | - | 8 | 1054 | 5243984 | ||
BC030665.1 | - | 7 | 983 | 5258865 | ||
CR456588.1 | - | 7 | 840 |