Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102724428 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320643.1 | NP_001307572.1 | 14 | 2599 | ||
XM_017028208.1 | XP_016883697.1 | 13 | 2451 | |||
GenBank mRNA | AK131076.1 | - | 13 | 2410 | ||
BC038504.1 | - | 14 | 2599 | 4831049 |
Gene Symbol: SIK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173354.3 | NP_775490.2 | 14 | 2593 | ||
XM_011529474.2 | XP_011527776.1 | 13 | 2450 | |||
GenBank mRNA | AB047786.1 | - | 14 | 2593 |