Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIEXF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014388.6 | NP_055203.4 | 12 | 2467 | ||
XM_006711275.3 | XP_006711338.1 | 10 | 2108 | |||
XM_011509418.2 | XP_011507720.1 | 12 | 2491 | |||
GenBank mRNA | AK123674.1 | - | 6 | 1273 | ||
BC022964.1 | - | 12 | 2428 | 5110673 | ||
BX648514.1 | - | 12 | 2380 | |||
CR749825.1 | - | 12 | 2404 |