Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181672.2 | NP_858058.1 | 13 | 1867 | ||
NM_181673.2 | NP_858059.1 | 13 | 1837 | |||
XM_017029907.1 | XP_016885396.1 | 11 | 1396 | |||
XM_017029908.1 | XP_016885397.1 | 11 | 4869 | |||
GenBank mRNA | AF223393.1 | - | 9 | 1504 | ||
AK313530.1 | - | 9 | 1783 | |||
AL050366.1 | - | 8 | 3094 | |||
AL833085.2 | - | 13 | 1847 | |||
BC014434.1 | - | 13 | 1768 | 4865031 | ||
BC038180.1 | - | 13 | 1800 | 5017795 | ||
BX537844.1 | - | 13 | 1825 | |||
U77413.1 | - | 9 | 1516 |