Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000911.3 | NP_000902.3 | 3 | 1664 | ||
GenBank mRNA | U07882.2 | - | 3 | 1661 |
Set Membership: |
G-protein coupled receptor |